Learn More
UBE2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529940
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: mouse brain. Predicted reactivity: Mouse (100%), Rat (93%), Zebrafish (100%), Xenopus laevis (100%), Chicken (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
UBE2A Polyclonal antibody specifically detects UBE2A in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
UBE2A | |
Polyclonal | |
Unconjugated | |
UBE2A | |
E2 ubiquitin-conjugating enzyme A; hhr6a; HR6A; Mhr6a; mp:zf637-2-000771; MRXS30; MRXSN; RAD6 homolog A; Rad6a; si:bz46j2.4; UBC2; ube2a; ube2a.L; Ubiquitin carrier protein A; ubiquitin conjugating enzyme E2 A; ubiquitin conjugating enzyme E2 A L homeolog; ubiquitin conjugating enzyme E2 A L homeolog; ubiquitin conjugating enzyme E2A L homeolog; ubiquitin conjugating enzyme E2A; ubiquitin-conjugating enzyme E2 A; ubiquitin-conjugating enzyme E2A; ubiquitin-conjugating enzyme E2A (RAD6 homolog); ubiquitin-conjugating enzyme E2A, RAD6 homolog; ubiquitin-conjugating enzyme HR6A; ubiquitin-protein ligase A; wu:fa01h11; XELAEV_18038882mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22209, 7319 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.3 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P49459, Q9Z255 | |
UBE2A | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 141 of Human UBE2A | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |