Learn More
UBE2J2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542351
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GIQLLNGHAP GAVPNLAGLQ QANRHHGLLG GALANLFVIV GFAAFAYTVK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%
UBE2J2 Polyclonal antibody specifically detects UBE2J2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
UBE2J2 | |
Polyclonal | |
Unconjugated | |
UBE2J2 | |
1200007B18Rik; 2400008G19Rik; 5730472G04Rik; AL022923; E2 ubiquitin-conjugating enzyme J2; NCUBE2; NCUBE-2; Non-canonical ubiquitin-conjugating enzyme 2; PRO2121; Ubc6; Ubc6p; Ubc6p homolog; UBE2J2; ubiquitin conjugating enzyme 6; ubiquitin conjugating enzyme E2 J2; ubiquitin conjugating enzyme E2, J2; ubiquitin-conjugating enzyme E2 J2; ubiquitin-conjugating enzyme E2, J2; ubiquitin-conjugating enzyme E2, J2 (UBC6 homolog, yeast); ubiquitin-conjugating enzyme E2, J2 homolog; ubiquitin-conjugating enzyme E2J 2; Unknown (protein for MGC:133997); yeast UBC6 homolog; zgc:162164 | |
Rabbit | |
Protein A | |
RUO | |
118424 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N2K1 | |
UBE2J2 | |
Synthetic peptide directed towards the C-terminal of human UBE2J2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |