Learn More
UBL4A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544975
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: EGEAQRLADS PPPQVWQLIS KVLARHFSAA DASRVLEQLQ RDYERSLSRL Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%; Yeast: 83%
UBL4A Polyclonal antibody specifically detects UBL4A in Human samples. It is validated for Western Blot.Specifications
UBL4A | |
Polyclonal | |
Unconjugated | |
UBL4A | |
DX254E; DXHXS254E; DXS254E; DXS254Eh; G6PD; GDX; GET5; housekeeping protein DXS254E; hypothetical protein LOC504533; MDY2; RGD1563983; TMA24; ubiquitin like 4A; ubiquitin-like 4; ubiquitin-like 4A; ubiquitin-like protein 4A; Ubiquitin-like protein GDX; UBL4; UBL4A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8266 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P11441 | |
UBL4A | |
Synthetic peptide directed towards the middle region of human UBL4A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |