missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK1 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen OSU00002G
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 500 μL of distilled water. Centrifuge to remove any insoluble material.
Involved in axon growth. Plays an essential role in neurite extension of cerebellar granule cells. Interacts with GABARAP and GABARAPL2.Specifications
ULK1 | |
Polyclonal | |
ULK1 | |
Unc-51-like kinase 1, serine/threonine-protein kinase Unc51.1 KIAA0722, ULK1, ATG1, UNC51, Unc51.1, unc-51-like kinase 1 (C. elegans), FLJ38455 | |
Sheep | |
Ammonium sulfate precipitation | |
RUO | |
22241, 360827, 8408 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Immunohistochemistry, Western Blot | |
Unconjugated | |
O75385, O70405 | |
ULK1 | |
A synthetic peptide from internal region of human ULK1 | |
500 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |