missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP39 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541515
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MSGRSKRESR GSTRGKRESE SRGSSGRVKR ERDREREPEA ASSRGSPVRV Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 100%; Rat: 100%
USP39 Polyclonal antibody specifically detects USP39 in Human samples. It is validated for Western Blot.Specifications
USP39 | |
Polyclonal | |
Unconjugated | |
USP39 | |
65K; AA408960; AI894154; CGI-21; D6Wsu157e; HSPC332; Inactive ubiquitin-specific peptidase 39; PRO2855; SAD1; SAD1 homolog; small nuclear ribonucleoprotein 65kDa (U4/U6.U5); SnRNP assembly defective 1 homolog; SNRNP65; U4/U6.U5 tri-snRNP-associated 65 kDa protein; U4/U6.U5 tri-snRNP-associated protein 2; ubiquitin specific peptidase 39; ubiquitin specific protease 39; Usp39 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10713 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q53GS9 | |
USP39 | |
Synthetic peptide directed towards the N-terminal of human USP39 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |