missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UXT Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518852
Additional Details : Weight : 0.01000kg
Description
UXT Polyclonal antibody specifically detects UXT in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
UXT | |
Polyclonal | |
Unconjugated | |
UXT | |
0910002B17Rik; androgen receptor trapped clone 27 protein; ART-27; HSPC024; Protein UXT; RP1-212G6.2; SKP2-associated alpha PFD 1; STAP1; ubiquitously expressed prefoldin like chaperone; ubiquitously expressed transcript; ubiquitously expressed transcript protein; ubiquitously-expressed transcript; ubiquitously-expressed, prefoldin-like chaperone; Unknown (protein for MGC:133475); UXT | |
Goat | |
Antigen affinity chromatography | |
RUO | |
8409 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9UBK9 | |
UXT | |
Synthetic peptide sequence (DHRDKVYEQLAKY) corresponding to the C-terminus amino acids of UXT | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |