Learn More
VCP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527323
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431, H1299, mouse brain, rat brain, A549, H1299, HCT-116. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (95%), Xenopus laevis (95%), Pig (100%), Chicken (99%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
VCP Polyclonal antibody specifically detects VCP in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
VCP | |
Polyclonal | |
Unconjugated | |
VCP | |
15S Mg(2+)-ATPase p97 subunit; 3110001E05; AAA ATPase p97; ALS14; CDC48; cell division cycle gene CDC48; CMT2Y; epididymis luminal protein 220; epididymis secretory protein Li 70; HEL-220; HEL-S-70; homolog of yeast cdc48; IBMPFD; IBMPFD1; Inv protein; or HEL-S-70; p97; p97 atpase; p97/VCP; Protein CDC48; si:ch211-113n10.2; TER ATPase; TERA; transitional endoplasmic reticulum ATPase; valosin containing protein; valosin-containing protein; Vcp; wu:fd16d05; wu:fj63d11; yeast Cdc48p homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116643, 269523, 7415 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P46462, P55072, Q01853 | |
VCP | |
Recombinant fragment contain a sequence corresponding to a region within amino acids 540 and 806 of VCP | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |