Learn More
VEGF Receptor 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527277
Additional Details : Weight : 0.01000kg
Description
PA5-27277 targets VEGF Receptor 3 in WB applications and shows reactivity with Human samples. The PA5-27277 immunogen is recombit protein fragment corresponding to a region within amino acids 1131 and 1352 of Human VEGF Receptor 3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
This gene encodes a tyrosine kinase receptor for vascular endothelial growth factors C and D. The protein is thought to be involved in lymphangiogenesis and maintece of the lymphatic endothelium. Mutations in this gene cause hereditary lymphedema type IA.Specifications
VEGF Receptor 3 | |
Polyclonal | |
Unconjugated | |
P35916 | |
FLT4 | |
Recombit protein fragment corresponding to a region within amino acids 1131 and 1352 of Human VEGF Receptor 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
FLT4 | |
FLT41, LMPH1A, PCL, VEGFR3, FLT-4, VEGFR-3, fms-like tyrosine kinase 4, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2324 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |