Learn More
VIPR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA3113
Additional Details : Weight : 0.01000kg
Description
PA3-113 detects VPAC1 Receptor in human samples. PA3-113 has been successfully used in Western blot, immunocytochemistry and immunohistochemistry (paraffin) procedures. By Western blot, this antibody detects an ∽55-70 kDa protein representing VPAC1 Receptor in human samples. In immunohistochemistry, VPAC1 Receptor immunoreactivity is predominantly localized at the level of the plasma membrane. PA3-113 immunizing peptide corresponds to amino acid residues 438-457 from human VPAC1 Receptor.
VIPR1 Polyclonal antibody specifically detects VIPR1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
VIPR1 | |
Polyclonal | |
Unconjugated | |
VIPR1 | |
AV071699; HVR1; II; Pacap receptor; PACAP type II receptor; PACAP-R2; PACAP-R-2; pituitary adenylate cyclase activating polypeptide receptor, type II; pituitary adenylate cyclase-activating polypeptide type II receptor; Pvr2; RATVASREC; RDC1; type 1 vasoactive intestinal peptide receptor; V1RG; VAPC1; vasoactive intestinal peptide receptor 1; vasoactive intestinal polypeptide receptor 1; Vasopressive intestinal peptide receptor; VASREC; VIP and PACAP receptor 1; VIP receptor 1; VIP receptor subtype 1; VIP receptor, type I; VIP1 receptor; VIPR; Vipr1; VIP-R1; VIP-R-1; VIRG; VPAC1; VPAC1 receptor; VPAC1R; VPCAP1R | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P32241, P97751 | |
VIPR1 | |
Synthetic peptide, KLH conjugated, corresponding to the residues T(438) R V S P G A R R S S S F Q A E V S L V(457) of human VPAC1 receptor. | |
RUO | |
22354, 7433 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |