Läs mer
Mettler Toledo™ Antistatic Kit, Universal
Produktkod. 10533744 Handla allt Mettler Toledo Produkter
Change view
:
For use with: Laboratory balances
Produktkod.
10533744 For use with: Laboratory balances
1 options
Produktkod. 10533744 Leverantör Mettler Toledo™ Leverantörsnummer 11107767
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Completely remove electrostatic charge from your samples and tare containers and avoid distortion of your weighing results. Mettler Toledo™ Antistatic Kit, Universal enables fast dosing processes and substantially reduces the risk of contamination.
  • Large electrode for discharging all kinds of samples and tare containers with dimensions of 15x25cm
  • Improves weighing accuracy
  • Enables fast dosing
  • Reduces contamination risk
  • Can be freely placed next to your balance
  • Includes universal power supply
TRUSTED_SUSTAINABILITY
Specifikationer
För användning med (utrustning) Laboratory Balance
För användning med (applikation) Discharging electrostatic charge
Inkluderar Universal power supply, Large U-Electrode
Storlek Large