Learn More
Invitrogen™ Anza™ 19 BglII
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand:  Invitrogen™ IVGN0198
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 15575700
  • 2850.00 SEK / 1 set
Estimated Shipment
14-12-2020
Add to basket
Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Description
Description
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity
Certifications
CoA
Specifications
Specifications
8,000U
Approved for shipment on Wet or Dry Ice
Not dam methylation-sensitive, dcm methylation-sensitive, Not CpG methylation-sensitive
Anza™ 10x Red Buffer, Anza™ 10x Buffer
37°C
800 µL Anza 19 BglII
2 mL Anza Buffer (10X)
2 mL Anza Red Buffer (10X)
Store at -20°C.
Anza™
Bgl II