Läs mer
Tocris Bioscience™ AP 18
GREENER_CHOICE
Produktkod. 18005942
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18005942 10 mg
18003982 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18005942 Leverantör Tocris Bioscience™ Leverantörsnummer 3296/10
Reversible TRPA1 channel blocker
AP 18 is a reversible TRPA1 channel blocker (IC50 values are 3.1 and 4.5 μM at human and mouse TRPA1 respectively). Blocks cinnamaldehyde-induced but not capsaicin-induced nociception and reverses mechanical hyperalgesia in vivo. Also blocks TRPA1 pore dilation (IC50 = 10.3 μM for the inhibition of Yo-Pro uptake).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55224-94-7
Molekylformel C11H12ClNO
Specifikationer
Kemiskt namn eller material AP 18
Kvantitet 10 mg
Formel vikt 209.67
Renhet >98%
Molekylvikt (g/mol) 209.673