Läs mer
Tocris Bioscience™ AP 1867
Beskrivning
AP 1867 is a selective binding ligand for the single point mutant of FKBP12F36V (IC50 = 1.8 nM). Functionalized with a carboxylic acid group at the meta-position to enable onward chemistry. The position of the carboxylic acid group represents an 'exit vector' allowing modification without interfering with the compound's binding ability.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (1R)-1-[3-(Carboxymethoxy)phenyl]-3-(3,4-dimethoxyphenyl)propyl (2S)-1-[(2S)-1-oxo-2-(3,4,5-trimethoxyphenyl)butyl]-2-piperidinecarboxylate |
| CAS | 195514-23-9 |
| Kvantitet | 50 mg |
| Mål | TAG Degradation Platforms |
| Molekylformel | C38H47NO11 |
| Renhet | 98% |