missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AP1S1 (Human) Recombinant Protein
Human AP1S1 full-length ORF ( AAH03561, 1 a.a. - 133 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001174-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MMRFMLLFSRQGKLRLRKWYLATSDKERKKMVRELMQVVLARKPKMCSFLEWRDLKVVYKRYASLYFCCAIEGQDNELITLELIHRYVELLDKYFGSVCELDIIFNFEKAYFILDEFLMGGDVQDTSTFPFSH
Specifications
AAH03561 | |
adaptor-related protein complex 1, sigma 1 subunit | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AP1S1 | |
GST |
1174 | |
Wheat germ expression system | |
10 μg | |
AP19, CLAPS1, FLJ92436, SIGMA1A, WUGSC:H_DJ0747G18.2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |