missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
AP2 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56332SS
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
AP2 alpha Polyclonal specifically detects AP2 alpha in Human, Mouse, Rat, Bovine, Canine, Primate, Sheep, Xenopus samples. It is validated for Western Blot, Dot Blot.
Specifikationer
| AP2 alpha | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen has 100% identity in human, mouse, rat, bovine, dog, monkey, sheep and Xenopus, and 93% in chicken. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Dot Blot | |
| 0.5 mg/mL | |
| Western Blot 1-2 ug/ml, Dot Blot | |
| NP_001035890 | |
| TFAP2A | |
| A portion of amino acids 100-150 of human AP-2 was used as the immunogen. | |
| 0.025 mg | |
| Cancer, Transcription Factors and Regulators | |
| 7020 | |
| Human, Mouse, Rat, Bovine, Canine, Primate, Sheep, Xenopus species | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering