missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15722-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Adaptins are components of the adaptor complexes which link clathrin to receptors in coated vesicles. Clathrin-associated protein complexes are believed to interact with the cytoplasmic tails of membrane proteins, leading to their selection and concentration. Alpha adaptin is a subunit of the plasma membrane adaptor. It binds polyphosphoinositides.Specifications
AP2A2 | |
Polyclonal | |
Unconjugated | |
AP2A2 | |
ADTAB, Alpha adaptin C, Alpha2 adaptin, AP 2 complex subunit alpha 2, AP2A2, CLAPA2, HIP 9, HIP9, Huntingtin yeast partner J, HYPJ, KIAA0899 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11772, 161, 81637 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O94973, P17427, P18484 | |
Ap2a2 | |
AP2A2 Fusion Protein Ag8442 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |