Läs mer
BD Biosciences™ APC Beads (CE/IVD)
Beskrivning
- BD CaliBRITE™ beads are designed for use with FACSComp™ or AutoCOMP™ software and the FACS™ family of flow cytometers (FACSCalibur™, FACSort™, FACScan™, and FACStrak™).
- Used to adjust instrument settings, set fluorescence compensation, and check instrument sensitivity.
- Daily use is recommended for monitoring instrument performance over time.
- CaliBRITE beads are for in vitro diagnostic use.
Specifikationer
Specifikationer
| Innehåll och lagring | Store BD Calibrite beads at 2°C–8°C and protected from direct light. Do not use after the expiration date shown on the label. Store Bead Dilution Buffer at 2°C–8°C. After dilution, BD Calibrite bead suspensions prepared in Bead Dilution Buffer are stable for 8 hours at 2°C–8°C or for 1 hour at 20°C–25°C. After dilution, BD Calibrite bead suspensions prepared in BD FACSFlow™ sheath fluid are stable for 8 hours at 2°C–8°C or, if PerCP beads are included, for 1 hour at 2°C–8°C. |
| Antal tester | 25 Tests |
| Kvantitet | 25 tester |