Läs mer
BD Biosciences™ APC Beads (CE/IVD)
Produktkod. 10433635
Change view
Kvantitet:
25 tester
Produktkod. Kvantitet
10433635 25 tester
1 options
Produktkod. 10433635 Leverantör BD Biosciences™ Leverantörsnummer 340487
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Used to adjust instrument settings, set fluorescence compensation, and check instrument sensitivity.
  • BD CaliBRITE™ beads are designed for use with FACSComp™ or AutoCOMP™ software and the FACS™ family of flow cytometers (FACSCalibur™, FACSort™, FACScan™, and FACStrak™).
  • Used to adjust instrument settings, set fluorescence compensation, and check instrument sensitivity.
  • Daily use is recommended for monitoring instrument performance over time.
  • CaliBRITE beads are for in vitro diagnostic use.
TRUSTED_SUSTAINABILITY
Specifikationer
Innehåll och lagring Store BD Calibrite beads at 2°C–8°C and protected from direct light. Do not use after the expiration date shown on the label. Store Bead Dilution Buffer at 2°C–8°C. After dilution, BD Calibrite bead suspensions prepared in Bead Dilution Buffer are stable for 8 hours at 2°C–8°C or for 1 hour at 20°C–25°C. After dilution, BD Calibrite bead suspensions prepared in BD FACSFlow™ sheath fluid are stable for 8 hours at 2°C–8°C or, if PerCP beads are included, for 1 hour at 2°C–8°C.
Antal tester 25 Tests
Kvantitet 25 tester