Learn More
R&D Systems™ APE1 Redox Inhibitor
Product Code. 18757663 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity Unit Size
18757663 5 mg 5mg
1 options
Product Code. 18757663 Supplier R&D Systems™ Supplier No. NBP149581
 Restricted Item
Beräknad leverans: 04-12-2026
A Novus product, part of R&D Systems. For use in research applications
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Quantity 5 mg
Product Line Novus
For Research Use Only