Läs mer
apo-Ipratropium Bromide, TRC
Produktkod. 30133673
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133673 10 mg
30133415 25 mg
30010307 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133673 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI77906910MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17812-46-3
Molekylformel C20H28NO2.Br
Molekylvikt (g/mol) 394.34
Synonym 1αH,5αH-Tropanium, 3α-hydroxy-8-isopropyl-, bromide, atropate (8CI), (3-endo)-8-Methyl-8-(1-methylethyl)-3-[(1-oxo-2-phenyl-2-propen-1-yl)oxy]-8-azoniabicyclo[3.2.1]octane bromide, (1R,3r,5S)-8-Methyl-8-(1-methylethyl)-3-[(2-phenylpropenoyl)oxy]-8-azoniabicyclo[3.2.1]octane bromide, (3-endo)-8-Isopropyl-8-methyl-3-[(2-phenylpropenoyl)oxy]-8-azoniabicyclo[3.2.1]octane bromide
IUPAC-namn [(1S,5R)-8-isopropyl-8-methyl-8-azoniabicyclo[3.2.1]octan-3-yl] 2-phenylprop-2-enoate;bromide
LEDER [Br-].CC(C)[N+]1(C)[C@@H]2CC[C@H]1CC(C2)OC(=O)C(=C)c3ccccc3
Specifikationer
Kemiskt namn eller material Ipratropium Bromide EP Impurity F
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C20H28NO2.BrH/c1-14(2)21(4)17-10-11-18(21)13-19(12-17)23-20(22)15(3)16-8-6-5-7-9-16;/h5-9,14,17-19H,3,10-13H2,1-2,4H3;1H/q+1;/p-1/t17-,18+,19+,21?;
Formel vikt 393.1303
Procent renhet >95%