missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ APOA1 (Human) Recombinant Protein
Human APOA1 full-length ORF ( AAH05380, 1 a.a. - 267 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00000335-P01.10ug
Weitere Details : Weight : 0.00010kg
Beschreibung
- Sequence: MKAAVLTLAVLFLTGSQARHFWQQDEPPQSPWDRVKDLATVYVDVLKDSGRDYVSQFEGSALGKQLNLKLLDNWDSVTSTFSKLREQLGPVTQEFWDNLEKETEGLRQEMSKDLEEVKAKVQPYLDDFQKKWQEEMELYRQKVEPLRAELQEGARQKLHELQEKLSPLGEEMRDRARAHVDALRTHLAPYSDELRQRLAARLEALKENGGARLAEYHAKATEHLSTLSEKAKPALEDLRQGLLPVLESFKVSFLSALEEYTKKLNTQ
Spezifikation
AAH05380 | |
apolipoprotein A-I | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APOA1 | |
GST |
335 | |
Wheat germ expression system | |
10 μg | |
MGC117399 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |