Läs mer
Apoatropine Hydrochloride, TRC
Produktkod. 30007035
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007035 250 mg
30011373 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007035 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA729050250MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5978-81-4
Molekylformel C17 H21 N O2 . Cl H
Molekylvikt (g/mol) 307.82
Synonym Apoatropine Hydrochloride, (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl 2-Phenylprop-2-enoate Hydrochloride, (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl 2-Phenylpropenoate Hydrochloride, Hyoscyamine Sulphate Imp. G (EP) as Hydrochloride, Atropine Imp. A (EP) as Hydrochloride
IUPAC-namn [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 2-phenylprop-2-enoate;hydrochloride
LEDER Cl.CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)C(=C)c3ccccc3
Specifikationer
Kemiskt namn eller material (1R,3r,5S)-8-Methyl-8-azabicyclo(3.2.1)oct-3-yl 2-Phenylpropenoate Hydrochloride (Apoatropine Hydrochloride)
Analyt- eller komponentnamn (1R,3r,5S)-8-Methyl-8-azabicyclo(3.2.1)oct-3-yl 2-Phenylpropenoate Hydrochloride (Apoatropine Hydrochloride)
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C17H21NO2.ClH/c1-12(13-6-4-3-5-7-13)17(19)20-16-10-14-8-9-15(11-16)18(14)2;/h3-7,14-16H,1,8-11H2,2H3;1H/t14-,15+,16+;
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 250 mg
Formel vikt 307.1339
Procent renhet >95%