missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27045-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein C gene family encodes four homologous proteins designated apoC-I to -IV, which specifically modulate the metabolism of triglyceride-rich lipoproteins. The human apoC-I gene maps to chromosome 19q13.2 and is expressed primarily in the liver where it is activated when monocytes differentiate into macrophages. The human apoC-II gene maps to chromosome 19q13.2 and encodes a 79 amino acid single chain protein that is a necessary cofactor for the activation of lipoprotein lipase, the enzyme that hydrolyzes triglycerides in plasma and transfers the fatty acids to tissues.Specifications
APOC2 | |
Polyclonal | |
Unconjugated | |
APOC2 | |
APC2, Apo CII, ApoC II, APOC2, apolipoprotein C II, Apolipoprotein C2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
344 | |
-20°C | |
Liquid |
Western Blot | |
0.67 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02655 | |
APOC2 | |
APOC2 Fusion Protein Ag24650 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |