missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein A II Mouse anti-Human, Clone: 1G1A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66773-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
At least 9 distinct polymorphic forms of apolipoproteins are known. The apolipoproteins act as stabilizers of the intact lipoprotein particles. Quantitative measurements of HDL, LDL, and VLDL particles of human serum are often used to estimate an individual′s relative risk of coronary heart disease. In addition, quantitative immunological measurements of certain apolipoproteins ( especially A-1 and B) have been suggested to be more accurate estimators of coronary heart disease that measurements of lipoprotein particles (especially HDL and LDL). The apolipoproteins are derived from human plasma by density gradient ultracentrifugation and HPLC.Specifications
Apolipoprotein A II | |
Monoclonal | |
0.8 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02652 | |
APOA2 | |
Apolipoprotein A II Fusion Protein Ag9863 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
1G1A2 | |
Unconjugated | |
APOA2 | |
Apo AII, ApoA II, APOA2, apolipoprotein A II, Apolipoprotein A2 | |
Mouse | |
Protein A | |
RUO | |
336 | |
-20°C | |
Liquid |