missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Apolipoprotein E/ApoE Antibody (WUE-4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60531AF700
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Apolipoprotein E/ApoE Monoclonal specifically detects Apolipoprotein E/ApoE in Human, Mouse, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifikationer
| Apolipoprotein E/ApoE | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571 | |
| Mouse | |
| 36 kDa | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 348 | |
| Human, Mouse, Rat (Negative) | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| WUE-4 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
| P02649 | |
| APOE | |
| Purified human ApoE [UniProt# P02649] | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody detects ApoE2, ApoE3 and ApoE4 (PMID 2466929). | |
| Store at 4C in the dark. | |
| IgG1 κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering