missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Apolipoprotein H/ApoH Antibody (APOH/3705) [PE], Novus Biologicals™

Product Code. 30025413 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
0.1 mL
missing translation for 'unitSize'
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30025413 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30025413 missing translation for 'mfr' Novus Biologicals™ missing translation for 'supplierNo' NBP326956PE

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Apolipoprotein H/ApoH Monoclonal antibody specifically detects Apolipoprotein H/ApoH in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein H/ApoH
Användningsområden Immunohistochemistry (Paraffin)
Klassificering Monoclonal
Klona APOH/3705
Konjugera PE
Formulering PBS
Gene Alias Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG
Värd art Mouse
Immunogen Human recombinant Apolipoprotein H/ApoH protein fragment (around aa 592-689) (exact sequence is proprietary)
Reningsmetod Protein A or G purified
Kvantitet 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Cholesterol Metabolism, Lipid and Metabolism
Primär eller sekundär Primary
Gen-ID (Entrez) 350
Målarter Human
Innehåll och lagring Store at 4°C in the dark.
Form Purified
Isotyp IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.