missing translation for 'onlineSavingsMsg'
Läs mer

Aprepitant N-Oxide, TRC

Produktkod. 30341730
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30341730 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30341730 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA72982525MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Aprepitant N-Oxide
CAS 172673-23-3
Molekylformel C23 H21 F7 N4 O4
Kvantitet 25 mg
Synonym [2R-[2α(R*),3α]]-5-[[2-[1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-oxido-4-morpholinyl]methyl]-1,2-dihydro-3H-1,2,4-triazol-3-one
InChI formel InChI=1S/C23H21F7N4O4/c1-12(14-8-15(22(25,26)27)10-16(9-14)23(28,29)30)38-20-19(13-2-4-17(24)5-3-13)34(36,6-7-37-20)11-18-31-21(35)33-32-18/h2-5,8-10,12,19-20H,6-7,11H2,1H3,(H2,31,32,33,35)/t12-,19+,20-,34?/m1/s1
LEDER C[C@@H](O[C@H]1OCC[N+]([O-])(CC2=NNC(=O)N2)[C@H]1c3ccc(F)cc3)c4cc(cc(c4)C(F)(F)F)C(F)(F)F
IUPAC-namn 3-[[(2R,3S)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-oxidomorpholin-4-ium-4-yl]methyl]-1,4-dihydro-1,2,4-triazol-5-one
Molekylvikt (g/mol) 550.426
Formel vikt 550.145
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.