missing translation for 'onlineSavingsMsg'
Learn More

AQP1 Mouse anti-Human, Porcine, Clone: 2E5D6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66805-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889015

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Aquaporin 1 is a 28kD integral membrane protein which was originally identified in red blood cells and renal proximal tubules. AQP1 is also expressed by the choroid plexus and various other tissues. It forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.
Specifications

Specifications

AQP1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P29972, Q6PQZ1
AQP1
AQP1 Fusion Protein Ag14093
20 μL
Primary
Human, Porcine
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
2E5D6
Unconjugated
AQP1
AQP 1, AQP CHIP, AQP1, Aquaporin 1, Aquaporin CHIP, CHIP28, CO, Urine water channel
Mouse
Protein G
RUO
358, 407773
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers