missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Aquaporin 1/AQP1 Antibody (1/A5F6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-749
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Aquaporin 1/AQP1 Monoclonal specifically detects Aquaporin 1/AQP1 in Human, Mouse, Rat, Rabbit, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifikationer
| Aquaporin 1/AQP1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:5000, ELISA 1:1000-1:20000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:500-1:1000 | |
| 28-kDa, AQP-1, AQP-CHIP, aquaporin 1 (channel-forming integral protein, 28kDa), aquaporin 1 (Colton blood group), Aquaporin-CHIP, CHIP2828kDa, CO blood group), CO, MGC26324 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 358 | |
| Human, Mouse, Rat, Rabbit, Zebrafish | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1/A5F6 | |
| Unconjugated | |
| P29975 | |
| AQP1 | |
| Synthetic peptide, corresponding to amino acids 249 - 269 of Rat Aquaporin 1. | |
| 0.05 mg | |
| Primary | |
| Antibody recognises an epitope within the cytoplasmic domain of the water-specific channel aquaporin 1. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering