missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Aquaporin 10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543530
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Peptide sequence: VGATVGTATY QLLVALHHPE GPEPAQDLVS AQHKASELET PASAQMLECK Sequence homology: Cow: 75%; Human: 100%; Rabbit: 75%; Rat: 79%.
AQP10 is a member of the aquaglyceroporin family of integral membrane proteins. Members of this family function as water-permeable channels in the epithelia of organs that absorb and excrete water. AQP10 was shown to function as a water-selective channel, and could also permeate neutral solutes such as glycerol and urea. This gene encodes a member of the aquaglyceroporin family of integral membrane proteins. Members of this family function as water-permeable channels in the epithelia of organs that absorb and excrete water. This protein was shown to function as a water-selective channel, and could also permeate neutral solutes such as glycerol and urea.
Specifikationer
| Aquaporin 10 | |
| Polyclonal | |
| Unconjugated | |
| AQP10 | |
| AQP10; AQP-10; AQPA_HUMAN; Aquaglyceroporin-10; aquaporin 10; aquaporin-10; Small intestine aquaporin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 89872 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96PS8 | |
| AQP10 | |
| Synthetic peptide directed towards the C-terminal of human AQP10 (aa 251-300). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering