Läs mer
(aR)-(1S,2S,3R,5S)-Pinanediol-1-amino-3-methylbutane-1-boronate Trifluoroacetic Acid Salt, TRC
Produktkod. 30409139
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30409139 250 mg
1 options
Produktkod. 30409139 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP458505250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R)-(S)-Pinanediol-1-amino-3-methylbutane-1-boronate trifluoroacetate salt
CAS 179324-87-9
Molekylformel C15 H28 B N O2 . C2 H F3 O2
Kvantitet 250 mg
Synonym 4,6-Methano-1,3,2-benzodioxaborole-2-methanamine','hexahydro-3a,8,8-trimethyl-α-(2-methylpropyl)-','(αR,3aS,4S,6S,7aR)-','2,2,2-trifluoroacetate (1:1), 4,6-Methano-1,3,2-benzodioxaborole-2-methanamine','hexahydro-3a,5,5-trimethyl-α-(2-methylpropyl)-','(αR,3aS,4S,6S,7aR)-','trifluoroacetate (9CI), 4,6-Methano-1,3,2-benzodioxaborole-2-methanamine','hexahydro-3a,5,5-trimethyl-α-(2-methylpropyl)-','[3aS-[2(S*),3aα,4β,6β,7aα]]-','trifluoroacetate, (R)-BoroLeu-(+)-Pinanediol Trifluoroacetate
InChI formel InChI=1S/C15H28BNO2.C2HF3O2/c1-9(2)6-13(17)16-18-12-8-10-7-11(14(10,3)4)15(12,5)19-16;3-2(4,5)1(6)7/h9-13H,6-8,17H2,1-5H3;(H,6,7)/t10-,11-,12+,13-,15-;/m0./s1
LEDER CC(C)C[C@H](N)B1O[C@@H]2C[C@@H]3C[C@@H](C3(C)C)[C@]2(C)O1.OC(=O)C(F)(F)F
Molekylvikt (g/mol) 379.22
Formel vikt 379.2142