Läs mer
AR-A014418, MedChemExpress
Produktkod. 16496727
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16496727 5 mg
16454018 10 mg
16400377 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16496727 Leverantör Medchem Express Leverantörsnummer HY10512
MedChemExpress AR-A014418 is a potent, selective and ATP-competitive GSK3β inhibitor (IC50=104 nM; Ki=38 nM).
AR-A014418 is a potent, selective and ATP-competitive GSK3β inhibitor (IC50=104 nM
Specifikationer
Kemiskt namn eller material AR-A014418
CAS 487021-52-3
Färg Earth Yellow
Anteckningar om renhetsgrad Research
Molekylformel C12H12N4O4S
Kvantitet 5 mg
Synonym AR 0133418 GSK 3β inhibitor VIII AR 014418
Löslighetsinformation DMSO : ≥ 100 mg/mL (324.35 mM)
LEDER O=C(NCC1=CC=C(OC)C=C1)NC2=NC=C([N+]([O-])=O)S2
Molekylvikt (g/mol) 308.31
Formel vikt 308.31
Procent renhet 99.49%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease