Läs mer
Tocris Bioscience™ AR-C 141990 hydrochloride
Beskrivning
AR-C 141990 hydrochloride is a monocarboxylate transporter (MCT1) inhibitor (pKi = 7.6. Exhibits approximately 10-fold selectivity for MCT1 over MCT2 and no significant activity against MCT3 or 4. Inhibits graft versus host response and prolongs cardiac graft survival in a rat model. Immunosuppressant. Also inhibits brain penetration of GHB analog HOCPCA.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 5-[[(3R)-3-Hydroxy-1-pyrrolidinyl]carbonyl]-3-methyl-1-(2-methylpropyl)-6-(4-quinolinylmethyl)thieno[2,3-d]pyrimidine-2,4(1H,3H)-dione |
| CAS | 2250019-94-2 |
| Analysprocentintervall | >98% |
| Kvantitet | 1 mg |
| Molekylformel | C26H28N4O4S.HCl |
| Formel vikt | Observed MW: 529.05 |