missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AR (Human) Recombinant Protein
Human AR partial ORF ( NP_000035, 221 a.a. - 320 a.a.) recombinant protein with GST-tag at N-terminal.
4055.00 SEK - 6150.00 SEK
Specifications
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
---|---|
Gene ID (Entrez) | 367 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16161761
|
Abnova™
H00000367-Q01.10UG |
10 μg |
4055.00 SEK
10µg |
Estimated Shipment: 28-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
16171761
|
Abnova™
H00000367-Q01.25UG |
25 μg |
6150.00 SEK
25µg |
Estimated Shipment: 28-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
- Sequence: SKDNYLGGTSTISDNAKELCKAVSVSMGLGVEALEHLSPGEQLRGDCMYAPLLGVPPAVRPTPCAPLAECKGSLLDDSAGKSTEDTAEYSPFKGGYTKGL
Specifications
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
367 |