missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AR Mouse anti-Human, Clone: 1F7C12, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | AR |
---|---|
Clone | 1F7C12 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888905
|
Proteintech
66747-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16878905
|
Proteintech
66747-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The androgen receptor (AR) is a member of the steroid-hormone receptor superfamily of nuclear receptors. The receptor is more than 90 kDa and has three major functional domains: the N-terminal domain, DNA-binding domain, and the androgen-binding domain. The androgen receptor is a ligand-activated transcription factor that binds active testosterone (T) and dihydrotestosterone (DHT). Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. The AR signaling pathway plays a key role in development and function of male reproductive organs, including the prostate and epididymis.AR also plays a role in nonreproductive organs, such as muscle, hair follicles, and brain.Androgen Receptor is a phosphoprotein, and also regulates mitogen-activated protein kinase (MAP kinase). The inhibition of the MEK1/2 pathway correlates directly with a change in phosphorylation state of the androgen receptor. Abnormalities in the AR signaling pathway have been linked to a number of diseases, including prostate cancer, Kennedy′s disease, and male infertility. Mutations in this gene are associated with complete androgen insensitivity (CAIS).Specifications
AR | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AIS, androgen receptor, AR, DHTR, Dihydrotestosterone receptor, HUMARA, HYSP1, KD, NR3C4, SBMA, SMAX1, TFM | |
AR | |
IgG2a | |
Protein A | |
Antibody |
1F7C12 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
P10275 | |
367 | |
AR Fusion Protein Ag17291 | |
Primary | |
-20°C | |
AR |