Läs mer
Arabic Acid, TRC
Produktkod. 30135922
Change view
Kvantitet:
2.5 g
5 g
25 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30135922 25 g -
30009483 5 g -
30006569 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135922 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA76421525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32609-14-6
Molekylformel C5 H10 O6
Molekylvikt (g/mol) 166.13
Synonym D-Arabinonic acid, Arabic acid, Arabonic acid, D-Arabinonate, Arabinoic acid
IUPAC-namn (2S,3R,4R)-2,3,4,5-tetrahydroxypentanoic acid
LEDER OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)O
Specifikationer
Kemiskt namn eller material D-Arabinic acid
Kvantitet 25 g
InChI formel InChI=1S/C5H10O6/c6-1-2(7)3(8)4(9)5(10)11/h2-4,6-9H,1H2,(H,10,11)/t2-,3-,4+/m1/s1
Formel vikt 166.0477