missing translation for 'onlineSavingsMsg'
Learn More

Arachidonic acid, >99%, MP Biomedicals™

Catalog Number p-4405716
Click to view available options
Quantity:
10 mg
50 mg
100 mg
This item is not returnable. View return policy
506-32-1
C20H32O2
304.474
YZXBAPSDXZZRGB-DOFZRALJSA-N
arachidonic acid, arachidonate, immunocytophyte, all-z-5,8,11,14-eicosatetraenoic acid, 5z,8z,11z,14z-icosa-5,8,11,14-tetraenoic acid, cis-5,8,11,14-eicosatetraenoic acid, 5,8,11,14-eicosatetraenoic acid, all-z, all-cis-5,8,11,14-eicosatetraenoic acid, unii-27yg812j1i, 5z,8z,11z,14z-eicosatetraenoic acid
444899
CHEBI:15843
(5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoic acid
CCCCCC=CCC=CCC=CCC=CCCCC(=O)O
This item is not returnable. View return policy
506-32-1
C20H32O2
304.474
YZXBAPSDXZZRGB-DOFZRALJSA-N
arachidonic acid, arachidonate, immunocytophyte, all-z-5,8,11,14-eicosatetraenoic acid, 5z,8z,11z,14z-icosa-5,8,11,14-tetraenoic acid, cis-5,8,11,14-eicosatetraenoic acid, 5,8,11,14-eicosatetraenoic acid, all-z, all-cis-5,8,11,14-eicosatetraenoic acid, unii-27yg812j1i, 5z,8z,11z,14z-eicosatetraenoic acid
444899
CHEBI:15843
(5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoic acid
CCCCCC=CCC=CCC=CCC=CCCCC(=O)O
Color Colorless
Density 0.922g/mL
Quantity 10 mg
Formula Weight 304.47
Percent Purity >99%
Physical Form Liquid
Chemical Name or Material Arachidonic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'