Läs mer
Arachidonic Acid Glycidyl Ester, TRC
Produktkod. 30345253
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30345253 50 mg
1 options
Produktkod. 30345253 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA76506050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Arachidonic Acid Glycidyl Ester
CAS 439146-24-4
Molekylformel C23 H36 O3
Kvantitet 50 mg
Synonym Glycidyl Arachidonate, (5Z,8Z,11Z,14Z)- 5,8,11,14-Eicosatetraenoic Acid Oxiranylmethyl Ester, Arachidonoyl-glycidyl Ester
InChI formel InChI=1S/C23H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23(24)26-21-22-20-25-22/h6-7,9-10,12-13,15-16,22H,2-5,8,11,14,17-21H2,1H3/b7-6-,10-9-,13-12-,16-15-
LEDER CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OCC1CO1
IUPAC-namn oxiran-2-ylmethyl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate
Molekylvikt (g/mol) 360.5301
Formel vikt 360.2664