Learn More
Arachidonic acid, MedChemExpress
Product Code. 16424318
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16424318 10 mg 10mg
16417037 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16424318 Supplier Medchem Express Supplier No. HY109590
 Restricted Item
Beräknad leverans: 30-11-2026
MedChemExpress Arachidonic acid is an essential fatty acid and a major constituent of biomembranes.
Arachidonic acid is an essential fatty acid and a major constituent of biomembranes.
Specifications
Chemical Name or Material Arachidonic acid
Physical Form Liquid
CAS 506-32-1
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H32O2
For Use With (Application) COVID-19-immunoregulation
Synonym Immunocytophyt
Solubility Information Ethanol : 100 mg/mL (328.44 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (164.22 mM)
SMILES CCCCC/C=C\C/C=C\C/C=C\C/C=C\CCCC(O)=O
Molecular Weight (g/mol) 304.47
Formula Weight 304.47
Percent Purity 98.0%
Grade Research