Läs mer
Arachidonic Acid N-Hydroxysuccinimidyl Ester, TRC
Produktkod. 30329709
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30329709 5 mg
1 options
Produktkod. 30329709 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7650105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Arachidonic Acid N-Hydroxysuccinimidyl Ester
CAS 187224-28-8
Molekylformel C24H35NO4
Kvantitet 5 mg
Synonym (all-Z)-1-[(1-Oxo-5,8,11,14-eicosatetraenyl)oxy]-2,5-pyrrolidinedione, 1-[[(5Z,8Z,11Z,14Z)-1-Oxo-5,8,11,14-eicosatetraenyl]oxy]-2,5-pyrrolidinedione, (5Z,8Z,11Z,14Z)-2,5-Dioxo-1-pyrrolidinyl Ester 5,8,11,14-Eicosatetraenoic Acid
InChI formel InChI=1S/C24H35NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-24(28)29-25-22(26)20-21-23(25)27/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-21H2,1H3/b7-6-,10-9-,13-12-,16-15-
LEDER O=C(ON1C(CCC1=O)=O)CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC
IUPAC-namn 2,5-dioxopyrrolidin-1-yl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate
Molekylvikt (g/mol) 401.55
Formel vikt 401.26