Läs mer
Aramite, TRC
Produktkod. 30341882
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30341882 10 mg
1 options
Produktkod. 30341882 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA76515310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Aramite
CAS 140-57-8
Anteckningar om renhetsgrad HPLC
Molekylformel C15 H23 Cl O4 S
Kvantitet 10 mg
Synonym Sulfurous acid, 2-(p-tert-butylphenoxy)-1-methylethyl 2-chloroethyl ester (6CI,8CI), Ethanol, 2-chloro-, 2-(p-tert-butylphenoxy)-1-methylethyl sulfite (8CI), 2-(p-tert-Butylphenoxy)-1-methylethyl 2-chloroethyl sulfite, 2-(p-tert-Butylphenoxy)isopropyl 2-chloroethyl sulfite, 88R, Aramit, Aramite, CES, NSC 404155, Niagaramite, Ortho-Mite, β-Chloroethyl-β'-(p-tert-butylphenoxy)-α'-methylethyl sulfite
InChI formel InChI=1S/C15H23ClO4S/c1-12(20-21(17)19-10-9-16)11-18-14-7-5-13(6-8-14)15(2,3)4/h5-8,12H,9-11H2,1-4H3
LEDER CC(COc1ccc(cc1)C(C)(C)C)OS(=O)OCCCl
IUPAC-namn 1-(4-tert-butylphenoxy)propan-2-yl 2-chloroethyl sulfite
Molekylvikt (g/mol) 334.86
Formel vikt 334.1006