Läs mer
Arbidol Sulfone, TRC
Produktkod. 30133784
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133784 100 mg
30004860 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133784 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA766010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 151455-32-2
Molekylformel C22H25BrN2O5S
Molekylvikt (g/mol) 509.41
Synonym 6-Bromo-4-[(dimethylamino)methyl]-5-hydroxy-1-methyl-2-[(phenylsulfonyl)methyl]-1H-indole-3-carboxylic Acid Ethyl Ester
IUPAC-namn ethyl 6-bromo-4-((dimethylamino)methyl)-5-hydroxy-1-methyl-2-((phenylsulfonyl)methyl)-1H-indole-3-carboxylate
LEDER BrC1=C(O)C(CN(C)C)=C2C(N(C)C(CS(=O)(C3=CC=CC=C3)=O)=C2C(OCC)=O)=C1
Specifikationer
Kemiskt namn eller material Arbidol Sulfone
Kvantitet 100 mg
InChI formel InChI=1S/C22H25BrN2O5S/c1-5-30-22(27)20-18(13-31(28,29)14-9-7-6-8-10-14)25(4)17-11-16(23)21(26)15(19(17)20)12-24(2)3/h6-11,26H,5,12-13H2,1-4H3
Formel vikt 509.42