Läs mer
Arbidol Sulfoxide, TRC
Produktkod. 30137831
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137831 100 mg
17983005 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137831 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA766005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 151455-33-3
Molekylformel C22H25BrN2O4S
Molekylvikt (g/mol) 493.41
Synonym 6-Bromo-4-[(dimethylamino)methyl]-5-hydroxy-1-methyl-2-[(phenylsulfinyl)methyl]-1H-indole-3-carboxylic Acid Ethyl Ester
IUPAC-namn ethyl 2-(benzenesulfinylmethyl)-6-bromo-4-[(dimethylamino)methyl]-5-hydroxy-1-methylindole-3-carboxylate
LEDER CCOC(=O)c1c(CS(=O)c2ccccc2)n(C)c3cc(Br)c(O)c(CN(C)C)c13
Specifikationer
Kemiskt namn eller material Arbidol Sulfoxide
Kvantitet 100 mg
InChI formel InChI=1S/C22H25BrN2O4S/c1-5-29-22(27)20-18(13-30(28)14-9-7-6-8-10-14)25(4)17-11-16(23)21(26)15(19(17)20)12-24(2)3/h6-11,26H,5,12-13H2,1-4H3
Formel vikt 492.0718