Läs mer
Tocris Bioscience™ Arbidol
Beskrivning
Arbidol is a broad spectrum antiviral. Displays activity against a range of respiratory and hepatitis viruses including influenza A, B and C, adenovirus, and hepatitis B and C. Increases stability of influenza viral hemagglutinins (HA) and prevents low-pH HA transition so inhibiting viral fusion. Also inhibits SARS-CoV-2 infection in vitro (IC50 = 4.11 μM).
Specifikationer
Specifikationer
| Kemiskt namn eller material | Ethyl 6-Bromo-4-((dimethylamino)methyl)-5-hydroxy-1-methyl-2-((phenylthio)methyl)-1 H-indole-3-carboxylate hydrochloride |
| CAS | 131707-23-8 |
| Kvantitet | 50 mg |
| Mål | Antiviral Compounds |
| Molekylformel | C22H25BrN2O3S.HCl |
| Renhet | 0.98 |