Läs mer
Tocris Bioscience™ ARCC 4 negative control
Beskrivning
ARCC 4 negative control is a negative control for ARCC 4 (Cat. No. 7254). Binds androgen receptors (AR) without inducing degradation.
Specifikationer
Specifikationer
| CAS | 2156588-26-8 |
| Kvantitet | 5 mg |
| Molekylformel | C53H56F3N7O7S2 |
| Renhet | 98% |