Läs mer
STEMCELL Technologies ArciTect™ High-Fidelity DNA Polymerase Kit
Beskrivning
- ArciTect™ High-Fidelity DNA Polymerase Kit is optimized for robust, high-fidelity DNA amplification (> 50X higher than Taq)
- It may be used in applications requiring ultra-low error rates, such as detection of genome editing with ArciTect™ T7 Endonuclease I (refer to the Technical Bulletin: Evaluation of Genome Editing), as well as sequencing, cloning/subcloning, synthetic biology, and SNP analysis
- ArciTect™ High-Fidelity DNA Polymerase is a fusion protein with a double-stranded DNA-binding domain and a Pyrococcus-like proofreading polymerase domain
Specifikationer
Specifikationer
| Beskrivning | Pluripotent Stem Cells |
| Kvantitet | 1 Kit |