missing translation for 'onlineSavingsMsg'
Learn More

ARD1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15670-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824944

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Displays alpha (N-terminal) acetyltransferase activity. Proposed alternative catalytic subunit of the N-terminal acetyltransferase A (NatA) complex.
Specifications

Specifications

ARD1B
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3UX61, Q4V8K3, Q9BSU3
ARD1 homolog B (S. cerevisiae), ARD1B, ARD2, hARD2, NAA11
Rabbit
IgG
150 μL
-20°C
Primary
289482, 84779, 97243
Western Blot
Unconjugated
NAA11
Liquid
Naa11
ARD1B Fusion Protein Ag8135
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers