Läs mer
Arecaidine Ethyl Ester Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Arecaidine Ethyl Ester Hydrochloride |
| CAS | 17210-50-3 |
| Molekylformel | C9 H15 N O2 . Cl H |
| Kvantitet | 50 mg |
| Synonym | 3-Pyridinecarboxylic acid, 1,2,5,6-tetrahydro-1-methyl-, ethyl ester, hydrochloride (1:1), 3-Pyridinecarboxylic acid, 1,2,5,6-tetrahydro-1-methyl-, ethyl ester, hydrochloride (9CI), Nicotinic acid, 1,2,5,6-tetrahydro-1-methyl-, ethyl ester, hydrochloride (8CI), Arecaidine ethyl ester hydrochloride |
| InChI formel | InChI=1S/C9H15NO2.ClH/c1-3-12-9(11)8-5-4-6-10(2)7-8;/h5H,3-4,6-7H2,1-2H3;1H |
| LEDER | Cl.CCOC(=O)C1=CCCN(C)C1 |
| IUPAC-namn | ethyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate;hydrochloride |
| Molekylvikt (g/mol) | 205.68 |
| Formel vikt | 205.087 |