Läs mer
Arecaidine Hydrobromide, TRC
Produktkod. 30133692
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133692 100 mg
30011729 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133692 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA767000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6013-57-6
Molekylformel C7 H11 N O2 . Br H
Molekylvikt (g/mol) 222.08
IUPAC-namn 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylic acid;hydrobromide
LEDER Br.CN1CCC=C(C1)C(=O)O
Specifikationer
Kemiskt namn eller material Arecaidine hydrobromide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C7H11NO2.BrH/c1-8-4-2-3-6(5-8)7(9)10;/h3H,2,4-5H2,1H3,(H,9,10);1H
Formel vikt 221.0051
Procent renhet >95%