missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ARF4 (Human) Recombinant Protein

Human ARF4 full-length ORF ( AAH03364, 1 a.a. - 180 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000378-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16181791

  • 5030.41 SEK / 25µg

Estimated Shipment: 11-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MGLTISSLFSRLFGKKQMRILMVGLDAAGKTTILYKLKLGEIVTTIPTIGFNVETVEYKNICFTVWDVGGQDRIRPLWKHYFQNTQGLIFVVDSNDRERIQEVADELQKMLLVDELRDAVLLLFANKQDLPNAMAISEMTDKLGLQSLRNRTWYVQATCATQGTGLYEGLDWLSNELSKR
Specifications

Specifications

AAH03364
ADP-ribosylation factor 4
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARF4
GST
378
Wheat germ expression system
25 μg
ARF2
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates