Läs mer
Arfolitixorin, TRC
Specifications
Specifications
| Kemiskt namn eller material | (6aR)-5,10-Methylenetetrahydrofolic Acid |
| CAS | 31690-11-6 |
| Molekylformel | C20 H23 N7 O6 |
| Kvantitet | 10 mg |
| Synonym | Arfolitixorin, L-Glutamic acid, N-[4-[(6aR)-3-amino-1,2,5,6,6a,7-hexahydro-1-oxoimidazo[1,5-f]pteridin-8(9H)-yl]benzoyl]-, Glutamic acid, N-[p-(3-amino-5,6,6a,7-tetrahydro-1-hydroxyimidazo[1,5-f]pteridin-8(9H)-yl)benzoyl]-, L-(+)- (8CI), L-Glutamic acid, N-[4-(3-amino-1,2,5,6,6a,7-hexahydro-1-oxoimidazo[1,5-f]pteridin-8(9H)-yl)benzoyl]-, (R)-, N-[4-[(6aR)-3-Amino-1,2,5,6,6a,7-hexahydro-1-oxoimidazo[1,5-f]pteridin-8(9H)-yl]benzoyl]-L-glutamic acid, (6R)-5,10-Methylene-5,6,7,8-tetrahydrofolic acid, (6R)-5,10-Methylenetetrahydrofolate, 5,10-Methylene-(6R)-tetrahydrofolic acid, Arfolitixorin, L-(+)-Methylenetetrahydrofolic acid, d-N5,N10-Methylene-L-tetrahydrofolic acid, (2S)-2-[4-[(6aR)-3-Amino-1-oxo-1,2,5,6,6a,7-hexahydroimidazo[1,5-f]pteridin-8(9H)-yl]benzamido]pentanedioic acid, (6aR)-5,10-Methylenetetrahydrofolic acid |
| InChI formel | InChI=1S/C20H23N7O6/c21-20-24-16-15(18(31)25-20)27-9-26(8-12(27)7-22-16)11-3-1-10(2-4-11)17(30)23-13(19(32)33)5-6-14(28)29/h1-4,12-13H,5-9H2,(H,23,30)(H,28,29)(H,32,33)(H4,21,22,24,25,31)/t12-,13+/m1/s1 |
| LEDER | NC1=NC(=O)C2=C(NC[C@@H]3CN(CN23)c4ccc(cc4)C(=O)N[C@@H](CCC(=O)O)C(=O)O)N1 |
| IUPAC-namn | (2S)-2-[[4-[(6aR)-3-amino-1-oxo-4,5,6,6a,7,9-hexahydroimidazo[1,5-f]pteridin-8-yl]benzoyl]amino]pentanedioic acid |
| Molekylvikt (g/mol) | 457.44 |
| Formel vikt | 457.171 |